Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101274.2 | NP_001094744.1 | 2 - 3 | 258 | 61 | |
GenBank mRNA | BC149691.1 | - | 2 - 3 | 259 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |