Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SALL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372185.1 | NP_001359114.1 | 1 - 2 | 307 | 118 | |
GenBank mRNA | BC149892.1 | - | 1 - 2 | 188 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |