Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOLL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102115.1 | NP_001095585.1 | 1 - 2 | 162 | 103 | |
NM_001168704.1 | NP_001162175.1 | 1 - 2 | 162 | 103 | ||
GenBank mRNA | BC149292.1 | - | 1 - 2 | 162 | 103 | |
EU050657.1 | - | 1 - 2 | 74 | 103 | ||
FJ623266.1 | - | 1 - 2 | 77 | 103 | ||
GU187434.1 | - | 1 - 2 | 162 | 103 | ||
GU187435.1 | - | 1 - 2 | 162 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |