Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102129.2 | NP_001095599.1 | 4 - 5 | 363 | 99 | |
GenBank mRNA | BC150053.1 | - | 4 - 5 | 364 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |