Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102129.2 | NP_001095599.1 | 8 - 9 | 686 | 73 | |
GenBank mRNA | BC150053.1 | - | 8 - 9 | 687 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |