Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102138.2 | NP_001095608.1 | 13 - 14 | 1879 | 85 | |
GenBank mRNA | BC150052.1 | - | 13 - 14 | 1881 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |