Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102189.2 | NP_001095659.2 | 2 - 3 | 278 | 54 | |
GenBank mRNA | AV598384.1 | - | 2 - 3 | 286 | 54 | |
BC149551.1 | - | 2 - 3 | 279 | 54 | ||
EE361823.1 | - | 2 - 3 | 268 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |