Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102246.1 | NP_001095716.1 | 1 - 2 | 374 | 63 | |
GenBank mRNA | BC126752.1 | - | 4 - 5 | 949 | 63 | |
BC149108.1 | - | 1 - 2 | 374 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |