Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102246.1 | NP_001095716.1 | 2 - 3 | 551 | 55 | |
GenBank mRNA | BC126752.1 | - | 5 - 6 | 1126 | 55 | |
BC149108.1 | - | 2 - 3 | 551 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |