Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102257.1 | NP_001095727.1 | 10 - 11 | 1172 | 63 | |
GenBank mRNA | BC149978.1 | - | 10 - 11 | 1172 | 63 | |
BC151441.1 | - | 10 - 11 | 1172 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |