Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102322.2 | 5 - 6 | 546 | 94 | ||
GenBank mRNA | BC149566.1 | - | 5 - 6 | 544 | 94 | |
KJ415257.1 | - | 3 - 4 | 315 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |