Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC27A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103273.1 | NP_001096743.1 | 4 - 5 | 1206 | 63 | |
GenBank mRNA | BC148156.1 | - | 4 - 5 | 1206 | 63 | |
KP735932.1 | - | 4 - 5 | 830 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |