Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104999.1 | NP_001098469.1 | 4 - 5 | 428 | 72 | |
GenBank mRNA | BC149395.1 | - | 4 - 5 | 428 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |