Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPACA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105007.1 | NP_001098477.1 | 3 - 4 | 571 | 142 | |
GenBank mRNA | BC149686.1 | - | 3 - 4 | 571 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |