Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105401.2 | NP_001098871.1 | 6 - 7 | 846 | 91 | |
GenBank mRNA | BC153239.1 | - | 6 - 7 | 847 | 91 | |
BT026286.1 | - | 5 - 6 | 743 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |