Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105402.2 | NP_001098872.1 | 5 - 6 | 578 | 113 | |
GenBank mRNA | BC134788.1 | - | 5 - 6 | 580 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |