Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105455.1 | NP_001098925.1 | 1 | 54 | 67 | |
GenBank mRNA | BC102562.1 | - | 1 | 54 | 67 | |
BC151476.1 | - | 1 | 51 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon |