Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110507.1 | NP_001103977.1 | 1 - 2 | 175 | 97 | |
GenBank mRNA | DN280438.1 | - | 1 - 2 | 122 | 97 | |
DN526838.1 | - | 1 - 2 | 185 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |