Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099206.1 | NP_001092676.1 | 2 - 3 | 992 | 86 | |
GenBank mRNA | BC142209.1 | - | 2 - 3 | 992 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |