Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAFA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099727.1 | NP_001093197.1 | 4 - 5 | 698 | 106 | |
GenBank mRNA | BC142202.1 | - | 4 - 5 | 698 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |