Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013400.2 | NP_001013418.2 | 1 - 2 | 177 | 53 | |
GenBank mRNA | BC147872.1 | - | 1 - 2 | 176 | 53 | |
DV822182.1 | - | 1 - 2 | 177 | 53 | ||
EE338630.1 | - | 1 - 2 | 177 | 53 | ||
EV615359.1 | - | 1 - 2 | 179 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |