Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101304.1 | NP_001094774.1 | 4 - 5 | 931 | 138 | |
GenBank mRNA | BC148030.1 | - | 4 - 5 | 931 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |