Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUZP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102336.1 | NP_001095806.1 | 7 - 8 | 725 | 107 | |
GenBank mRNA | BC149520.1 | - | 7 - 8 | 725 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |