Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102340.1 | NP_001095810.1 | 3 - 4 | 467 | 97 | |
GenBank mRNA | BC148020.1 | - | 3 - 4 | 467 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |