Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102351.1 | NP_001095821.1 | 8 - 9 | 1732 | 80 | |
GenBank mRNA | BC148938.1 | - | 8 - 9 | 1732 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |