Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERCC6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102530.2 | NP_001096000.1 | 1 - 2 | 112 | 87 | |
GenBank mRNA | BC149963.1 | - | 1 - 2 | 113 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |