Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOBTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103104.1 | NP_001096574.1 | 7 - 8 | 1871 | 67 | |
GenBank mRNA | BC148035.1 | - | 7 - 8 | 1871 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |