Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103106.2 | NP_001096576.1 | 6 - 7 | 1738 | 80 | |
GenBank mRNA | BC150136.1 | - | 6 - 7 | 1739 | 80 | |
BC151633.1 | - | 7 - 8 | 1448 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |