Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105019.1 | NP_001098489.1 | 7 - 8 | 894 | 79 | |
GenBank mRNA | BC142236.1 | - | 7 - 8 | 894 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |