Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105030.2 | NP_001098500.1 | 4 - 5 | 951 | 66 | |
GenBank mRNA | BC150042.1 | - | 4 - 5 | 952 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |