Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105042.1 | NP_001098512.1 | 2 - 3 | 767 | 100 | |
GenBank mRNA | BC140551.1 | - | 2 - 3 | 767 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |