Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPIND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105046.2 | NP_001098516.1 | 3 - 4 | 1240 | 53 | |
GenBank mRNA | BC149414.1 | - | 3 - 4 | 1241 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |