Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105479.1 | NP_001098949.1 | 1 - 2 | 102 | 68 | |
GenBank mRNA | BC148959.1 | - | 1 - 2 | 102 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |