Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105479.1 | NP_001098949.1 | 4 - 5 | 425 | 108 | |
GenBank mRNA | BC148959.1 | - | 4 - 5 | 425 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |