Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFXANK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105488.1 | NP_001098958.1 | 2 - 3 | 310 | 72 | |
GenBank mRNA | BC148968.1 | - | 2 - 3 | 310 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |