Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IZUMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105642.1 | NP_001099112.1 | 5 - 6 | 721 | 73 | |
GenBank mRNA | BC147884.1 | - | 5 - 6 | 721 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |