Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014902.4 | NP_001014902.1 | 2 - 3 | 360 | 61 | |
GenBank mRNA | BC104565.1 | - | 2 - 3 | 358 | 61 | |
EV620976.1 | - | 2 - 3 | 360 | 61 | ||
EV632625.1 | - | 2 - 3 | 367 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Inventoried Probe spans exons |