Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099396.1 | NP_001092866.1 | 1 - 2 | 164 | 153 | |
GenBank mRNA | BC146030.1 | - | 1 - 2 | 164 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |