Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078149.1 | NP_001071617.1 | 6 - 7 | 604 | 143 | |
GenBank mRNA | BC102540.1 | - | 6 - 7 | 604 | 143 | |
U49179.1 | - | 6 - 7 | 595 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |