Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP299 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076389.2 | NP_001069857.1 | 6 | 746 | 134 | |
GenBank mRNA | BC110236.1 | - | 6 | 747 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |