Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KTI12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080737.1 | NP_001074206.1 | 1 | 1445 | 160 | |
GenBank mRNA | BC118297.1 | - | 1 | 1445 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |