Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098465.1 | NP_001091935.1 | 4 | 1148 | 113 | |
GenBank mRNA | BC134759.1 | - | 4 | 1148 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |