Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSKU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103268.1 | NP_001096738.1 | 2 | 2451 | 84 | |
GenBank mRNA | BC151665.1 | - | 2 | 2451 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |