Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205781.2 | NP_991350.1 | 1 | 2444 | 79 | |
GenBank mRNA | BC140513.1 | - | 1 | 2444 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |