Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATHL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174832.3 | NP_777257.1 | 4 | 441 | 117 | |
GenBank mRNA | X97608.1 | - | 3 - 4 | 445 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |