Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130756.1 | NP_001124228.1 | 2 - 3 | 540 | 72 | |
GenBank mRNA | BC123809.1 | - | 2 - 3 | 540 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |