Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNHG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038147.1 | 4 - 5 | 933 | 62 | ||
NR_038148.1 | 3 - 4 | 888 | 62 | |||
GenBank mRNA | EH132547.1 | - | 3 - 4 | 497 | 62 | |
EU848421.1 | - | 4 - 5 | 934 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |