Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC24A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144094.2 | NP_001137566.2 | 1 - 2 | 981 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |