Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145428.1 | NP_001138900.1 | 2 - 3 | 270 | 66 | |
GenBank mRNA | DV906549.1 | - | 2 - 3 | 261 | 66 | |
EE215471.1 | - | 2 - 3 | 272 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |