Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001374469.1 | NP_001361398.1 | 1 - 2 | 269 | 70 | |
GenBank mRNA | BC150095.1 | - | 1 - 2 | 260 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |